Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAGTCCACATGTGATTGACTCCT[C/T]CTTCTATTGCTTTGTACAGGGCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613222 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNPDA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GNPDA2 - glucosamine-6-phosphate deaminase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270880.1 | 747 | Missense Mutation | GAA,GGA | E,G 184 | NP_001257809.1 | |
NM_001270881.1 | 747 | Missense Mutation | GAA,GGA | E,G 148 | NP_001257810.1 | |
NM_138335.2 | 747 | Missense Mutation | GAA,GGA | E,G 218 | NP_612208.1 |
GUF1 - GUF1 homolog, GTPase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |