Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGACGTGAAGCCCACCAAGGCGCA[C/T]AGCCTCAACTCCGGTGCCTGCCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608201 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK5RAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK5RAP2 - CDK5 regulatory subunit associated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011649.2 | 6031 | UTR 3 | NP_001011649.1 | |||
NM_001272039.1 | 6031 | UTR 3 | NP_001258968.1 | |||
NM_018249.5 | 6031 | UTR 3 | NP_060719.4 | |||
XM_006717182.1 | 6031 | UTR 3 | XP_006717245.1 | |||
XM_006717185.1 | 6031 | UTR 3 | XP_006717248.1 | |||
XM_011518860.1 | 6031 | UTR 3 | XP_011517162.1 | |||
XM_017014921.1 | 6031 | UTR 3 | XP_016870410.1 | |||
XM_017014922.1 | 6031 | UTR 3 | XP_016870411.1 | |||
XM_017014923.1 | 6031 | UTR 3 | XP_016870412.1 | |||
XM_017014924.1 | 6031 | UTR 3 | XP_016870413.1 |