Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATTGTAATATAAAGACCATTAC[A/G]TGACAAAGAAGAAATTGGTTACAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602136 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PEX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PEX1 - peroxisomal biogenesis factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM48 - RNA binding motif protein 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032120.3 | 562 | Missense Mutation | ATG,GTG | M,V 153 | NP_115496.2 | |
XM_005250636.4 | 562 | Missense Mutation | ATG,GTG | M,V 153 | XP_005250693.1 | |
XM_006716149.3 | 562 | Missense Mutation | ATG,GTG | M,V 153 | XP_006716212.1 | |
XM_006716150.3 | 562 | Missense Mutation | ATG,GTG | M,V 153 | XP_006716213.1 | |
XM_017012709.1 | 562 | UTR 5 | XP_016868198.1 |