Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAACCAGGGCTTCCTAGATGTGG[A/T]AGCTGCTTTGCGCTGGGTGCAAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605279 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CES3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CES3 - carboxylesterase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185176.1 | 687 | Intron | NP_001172105.1 | |||
NM_001185177.1 | 687 | Missense Mutation | GAA,GTA | E,V 203 | NP_001172106.1 | |
NM_024922.5 | 687 | Missense Mutation | GAA,GTA | E,V 203 | NP_079198.2 | |
XM_011522975.2 | 687 | Intron | XP_011521277.1 | |||
XM_011522976.2 | 687 | Missense Mutation | GAA,GTA | E,V 203 | XP_011521278.1 |