Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGGATCACACTGGAGAGAAACC[A/C]TATGCTTGTAAAGTCTGTGGAAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 194543 | ||||||||||||||||||||
Literature Links: |
ZNF69 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF69 - zinc finger protein 69 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321163.1 | 731 | Intron | NP_001308092.1 | |||
NM_021915.3 | 731 | Intron | NP_068734.1 | |||
XM_006722873.3 | 731 | Intron | XP_006722936.1 | |||
XM_006722874.3 | 731 | Intron | XP_006722937.1 | |||
XM_017027231.1 | 731 | Intron | XP_016882720.1 |
ZNF700 - zinc finger protein 700 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271848.1 | 731 | Silent Mutation | CCA,CCC | P,P 196 | NP_001258777.1 | |
NM_144566.2 | 731 | Silent Mutation | CCA,CCC | P,P 193 | NP_653167.1 |