Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGATGACTCCTTAAGTGGTTCATC[A/G]TCTGCATCTTCGTGTGAACCAGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF512 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF512 - zinc finger protein 512 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271286.1 | 309 | Silent Mutation | TCA,TCG | S,S 55 | NP_001258215.1 | |
NM_001271287.1 | 309 | UTR 5 | NP_001258216.1 | |||
NM_001271288.1 | 309 | UTR 5 | NP_001258217.1 | |||
NM_001271289.1 | 309 | UTR 5 | NP_001258218.1 | |||
NM_001271318.1 | 309 | UTR 5 | NP_001258247.1 | |||
NM_032434.3 | 309 | Silent Mutation | TCA,TCG | S,S 56 | NP_115810.2 |