Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGGAGGCTGTGAAAACAACTGG[C/T]CTTTCCCAGGAACTCCAGGCAAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614518 MIM: 602136 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GATAD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GATAD1 - GATA zinc finger domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021167.4 | 3634 | Intron | NP_066990.3 |
PEX1 - peroxisomal biogenesis factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000466.2 | 3634 | Missense Mutation | GAC,GGC | D,G 1168 | NP_000457.1 | |
NM_001282677.1 | 3634 | Missense Mutation | GAC,GGC | D,G 1111 | NP_001269606.1 | |
NM_001282678.1 | 3634 | Missense Mutation | GAC,GGC | D,G 960 | NP_001269607.1 | |
XM_017012319.1 | 3634 | Missense Mutation | GAC,GGC | D,G 585 | XP_016867808.1 |