Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACTGTGGATGATGGCTGCAGACA[G/T]CCCGAGAGTGACCACTCCGCTGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZSCAN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZSCAN2 - zinc finger and SCAN domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007072.1 | 243 | Missense Mutation | AGC,ATC | S,I 6 | NP_001007073.1 | |
NM_017894.5 | 243 | Missense Mutation | AGC,ATC | S,I 6 | NP_060364.4 | |
NM_181877.3 | 243 | Missense Mutation | AGC,ATC | S,I 6 | NP_870992.2 | |
XM_017022392.1 | 243 | Missense Mutation | AGC,ATC | S,I 6 | XP_016877881.1 | |
XM_017022393.1 | 243 | Missense Mutation | AGC,ATC | S,I 6 | XP_016877882.1 |