Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGAAGTTGGGAACTATTTCGGTG[G/T]GCAGGATGCGGTACCGTTTGGGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600921 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGF9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGF9 - fibroblast growth factor 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002010.2 | 875 | Missense Mutation | GGG,GTG | G,V 13 | NP_002001.1 | |
XM_011534996.2 | 875 | Intron | XP_011533298.1 |