Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCATACTGGATGAGGATGTCTTT[A/G]ATCTCCTTCTTGGAAGCCTCATCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611893 MIM: 603675 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC723805 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC723805 - interleukin-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLEKHG2 - pleckstrin homology and RhoGEF domain containing G2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS16 - ribosomal protein S16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001020.5 | 398 | Silent Mutation | ATC,ATT | I,I 108 | NP_001011.1 | |
NM_001321111.1 | 398 | Silent Mutation | ATC,ATT | I,I 91 | NP_001308040.1 | |
XM_005259137.2 | 398 | Intron | XP_005259194.1 |