Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCTTGGGGATGCCCAGTGATAT[A/G]CACGTTGAAGCAGTAATGTAAGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 243305 MIM: 616717 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INVS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INVS - inversin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318381.1 | 2444 | Intron | NP_001305310.1 | |||
NM_001318382.1 | 2444 | Intron | NP_001305311.1 | |||
NM_014425.4 | 2444 | Intron | NP_055240.2 |
TEX10 - testis expressed 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161584.1 | 2444 | Missense Mutation | CAT,TAT | H,Y 898 | NP_001155056.1 | |
NM_017746.3 | 2444 | Missense Mutation | CAT,TAT | H,Y 914 | NP_060216.2 | |
XM_011518798.1 | 2444 | Missense Mutation | CAT,TAT | H,Y 789 | XP_011517100.1 | |
XM_017014848.1 | 2444 | Missense Mutation | CAT,TAT | H,Y 559 | XP_016870337.1 |