Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATATATAGTATACTTCACTTTCT[C/T]TACATGTTGTTCTGCACCTGCCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616075 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEFB121 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DEFB121 - defensin beta 121 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011878.2 | 500 | Missense Mutation | AAA,AGA | K,R 35 | NP_001011878.1 | |
NM_001171832.1 | 500 | Missense Mutation | AAA,AGA | K,R 23 | NP_001165303.1 | |
XM_005260383.3 | 500 | Missense Mutation | AAA,AGA | K,R 23 | XP_005260440.1 |