Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACTACCAGATATTCAGAGATTTC[A/G]AAGGCTAGTAGAGTCTTCAGTGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 313650 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TAF1 - TATA-box binding protein associated factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286074.1 | Intron | NP_001273003.1 | ||||
NM_004606.4 | Intron | NP_004597.2 | ||||
NM_138923.3 | Intron | NP_620278.1 | ||||
XM_005262295.1 | Intron | XP_005262352.1 | ||||
XM_005262296.1 | Intron | XP_005262353.1 | ||||
XM_005262297.4 | Intron | XP_005262354.1 | ||||
XM_005262300.2 | Intron | XP_005262357.1 | ||||
XM_006724682.3 | Intron | XP_006724745.1 |