Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGCCCCAGGAGCTCCAGGTAGC[C/T]CGCATCCTCATGGTAGCCTCCCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603954 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLDN25 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLDN25 - claudin 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101389.1 | 240 | Silent Mutation | GCC,GCT | A,A 80 | NP_001094859.1 |
LOC390251 - SH3 domain containing GRB2 like 1 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZW10 - zw10 kinetochore protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |