Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGCAGGTGGAGATGGTCTAGTC[C/T]TCCTGGGGCCTCTCCACAGTGAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603020 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AFG3L1P PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AFG3L1P - AFG3 like matrix AAA peptidase subunit 1, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DBNDD1 - dysbindin domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042610.2 | 605 | Silent Mutation | GAA,GAG | E,E 157 | NP_001036075.1 | |
NM_001288708.1 | 605 | Silent Mutation | GAA,GAG | E,E 157 | NP_001275637.1 | |
NM_001288709.1 | 605 | Silent Mutation | GAA,GAG | E,E 277 | NP_001275638.1 | |
NM_024043.3 | 605 | Silent Mutation | GAA,GAG | E,E 177 | NP_076948.2 |