Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGGGAGCTGAGGAGGGCCAAGA[T/C]GAGTCTAGGGCCTTGGTGGGCGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182282 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNORD119 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNORD119 - small nucleolar RNA, C/D box 119 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNRPB - small nuclear ribonucleoprotein polypeptides B and B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003091.3 | 1037 | UTR 3 | NP_003082.1 | |||
NM_198216.1 | 1037 | UTR 3 | NP_937859.1 |