Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTCTGAAACGCTTTCTTAGCAAA[A/G]CCAGGAGCGTCCCACCCATTATCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612309 MIM: 173610 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
F5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
F5 - coagulation factor V | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SELP - selectin P | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003005.3 | 2254 | Missense Mutation | GCT,GTT | A,V 793 | NP_002996.2 | |
XM_005245435.1 | 2254 | Missense Mutation | GCT,GTT | A,V 793 | XP_005245492.1 | |
XM_005245436.3 | 2254 | Missense Mutation | GCT,GTT | A,V 793 | XP_005245493.1 | |
XM_005245438.1 | 2254 | Intron | XP_005245495.1 | |||
XM_005245439.1 | 2254 | Intron | XP_005245496.1 | |||
XM_005245440.1 | 2254 | Missense Mutation | GCT,GTT | A,V 731 | XP_005245497.1 |