Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTGTGCCTTTCACCTGGGTGACT[A/T]CAAGAGAGCTCTGGAGGTTAGTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTC26 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTC26 - tetratricopeptide repeat domain 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144920.2 | 261 | Missense Mutation | TAC,TTC | Y,F 73 | NP_001138392.1 | |
NM_001144923.2 | 261 | Intron | NP_001138395.1 | |||
NM_001287512.1 | 261 | Missense Mutation | TAC,TTC | Y,F 73 | NP_001274441.1 | |
NM_001287513.1 | 261 | UTR 5 | NP_001274442.1 | |||
NM_001318333.1 | 261 | UTR 5 | NP_001305262.1 | |||
NM_001321740.1 | 261 | Missense Mutation | TAC,TTC | Y,F 73 | NP_001308669.1 | |
NM_001321741.1 | 261 | Missense Mutation | TAC,TTC | Y,F 73 | NP_001308670.1 | |
NM_001321742.1 | 261 | Missense Mutation | TAC,TTC | Y,F 73 | NP_001308671.1 | |
NM_024926.3 | 261 | Missense Mutation | TAC,TTC | Y,F 73 | NP_079202.2 |