Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAACGAGAGGACTCTTATGCCCAA[A/G]GCGCGTGAGCGAAGCCGGAGACTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616196 MIM: 610815 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCAF13 - DDB1 and CUL4 associated factor 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015420.6 | 260 | Missense Mutation | AAG,AGG | K,R 29 | NP_056235.4 |
SLC25A32 - solute carrier family 25 member 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030780.4 | 260 | UTR 5 | NP_110407.2 | |||
XM_017013877.1 | 260 | UTR 5 | XP_016869366.1 |