Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCACTTGGATTGATCAGGTGTGT[C/G]GGGTACTGCAGCCCCTGCTCGCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605435 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRKD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRKD1 - protein kinase D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002742.2 | 5204 | Silent Mutation | CCC,CCG | P,P 881 | NP_002733.2 | |
XM_005267859.1 | 5204 | Silent Mutation | CCC,CCG | P,P 889 | XP_005267916.1 | |
XM_011536964.2 | 5204 | Silent Mutation | CCC,CCG | P,P 813 | XP_011535266.1 | |
XM_011536965.2 | 5204 | Silent Mutation | CCC,CCG | P,P 793 | XP_011535267.1 | |
XM_017021461.1 | 5204 | Silent Mutation | CCC,CCG | P,P 793 | XP_016876950.1 | |
XM_017021462.1 | 5204 | Silent Mutation | CCC,CCG | P,P 716 | XP_016876951.1 |