Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAATGTCACGACTTGGTCTTCCC[G/T]AGGAGCCAGTCAGGTAAAGGCTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609231 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDT7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUDT7 - nudix hydrolase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105663.2 | 113 | Missense Mutation | GAG,TAG | E,* 8 | NP_001099133.1 | |
NM_001243657.1 | 113 | Missense Mutation | GAG,TAG | E,* 8 | NP_001230586.1 | |
NM_001243660.1 | 113 | Missense Mutation | GAG,TAG | E,* 8 | NP_001230589.1 | |
NM_001243661.1 | 113 | Missense Mutation | GAG,TAG | E,* 8 | NP_001230590.1 | |
XM_011523032.2 | 113 | Missense Mutation | GAG,TAG | E,* 8 | XP_011521334.1 | |
XM_011523033.2 | 113 | Missense Mutation | GAG,TAG | E,* 8 | XP_011521335.1 | |
XM_017023170.1 | 113 | Missense Mutation | GAG,TAG | E,* 8 | XP_016878659.1 |