Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGGATCTGGGCCTGTTTGCGCTC[A/T]ATCAGCTTCTTGATGATCTGAGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608588 MIM: 608344 MIM: 602301 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHX58 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHX58 - DEXH-box helicase 58 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HSPB9 - heat shock protein family B (small) member 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KAT2A - lysine acetyltransferase 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021078.2 | 2129 | Silent Mutation | ATA,ATT | I,I 679 | NP_066564.2 | |
XM_006721817.3 | 2129 | Silent Mutation | ATA,ATT | I,I 680 | XP_006721880.1 | |
XM_006721818.3 | 2129 | Silent Mutation | ATA,ATT | I,I 319 | XP_006721881.1 | |
XM_017024448.1 | 2129 | Silent Mutation | ATA,ATT | I,I 318 | XP_016879937.1 |