Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAGCAGAGAGCAAAGGCAGCTCA[C/T]CTCACGAAACTTCCAGGAGCGCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603319 MIM: 606920 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANXA9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANXA9 - annexin A9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003568.2 | 425 | Missense Mutation | ACC,ATC | T,I 79 | NP_003559.2 | |
XM_011510058.2 | 425 | Missense Mutation | ACC,ATC | T,I 79 | XP_011508360.1 |
CERS2 - ceramide synthase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |