Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCTGCACCCCTGCACTGTCCTC[A/G]GTGGTGGTGTGGAGGGCGGGGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614045 MIM: 610933 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM129B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM129B - family with sequence similarity 129 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035534.2 | 2239 | Silent Mutation | ACC,ACT | T,T 723 | NP_001030611.1 | |
NM_022833.3 | 2239 | Silent Mutation | ACC,ACT | T,T 736 | NP_073744.2 | |
XM_005252135.2 | 2239 | Silent Mutation | ACC,ACT | T,T 761 | XP_005252192.2 | |
XM_011518925.1 | 2239 | Silent Mutation | ACC,ACT | T,T 766 | XP_011517227.1 |
LRSAM1 - leucine rich repeat and sterile alpha motif containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |