Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGGTGGGGAACGCCAAGCACAA[C/T]GTGCATGTCATGAACATCTCCACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300512 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDR13 - WD repeat domain 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166426.1 | 1010 | Silent Mutation | AAC,AAT | N,N 192 | NP_001159898.1 | |
NM_017883.4 | 1010 | Silent Mutation | AAC,AAT | N,N 284 | NP_060353.2 | |
XM_006724539.1 | 1010 | Silent Mutation | AAC,AAT | N,N 284 | XP_006724602.1 | |
XM_006724541.3 | 1010 | Silent Mutation | AAC,AAT | N,N 192 | XP_006724604.1 | |
XM_011543947.2 | 1010 | Silent Mutation | AAC,AAT | N,N 284 | XP_011542249.1 |