Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTCCATCTTGTCATTACTTGAGG[G/T]CTTGTGTTTCTTTGGTAATTCCGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614762 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLYATL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLYATL2 - glycine-N-acyltransferase like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145016.3 | 743 | Missense Mutation | ACC,CCC | T,P 143 | NP_659453.3 | |
XM_011544809.2 | 743 | Missense Mutation | ACC,CCC | T,P 102 | XP_011543111.1 | |
XM_017017337.1 | 743 | Missense Mutation | ACC,CCC | T,P 143 | XP_016872826.1 | |
XM_017017338.1 | 743 | Missense Mutation | ACC,CCC | T,P 143 | XP_016872827.1 |