Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGATGATGAATGCTGAAAGTAC[A/C]ACCTGAACAGCAAAGGTCCATGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPAN19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TSPAN19 - tetraspanin 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100917.1 | 342 | Silent Mutation | GTG,GTT | V,V 101 | NP_001094387.1 | |
XM_005268669.4 | 342 | Silent Mutation | GTG,GTT | V,V 101 | XP_005268726.1 | |
XM_017018862.1 | 342 | Silent Mutation | GTG,GTT | V,V 101 | XP_016874351.1 | |
XM_017018863.1 | 342 | Silent Mutation | GTG,GTT | V,V 101 | XP_016874352.1 | |
XM_017018864.1 | 342 | Silent Mutation | GTG,GTT | V,V 101 | XP_016874353.1 |