Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAACCTTCATAAGCTTGACAATGT[A/G]GAATTGTATTTCTTAAAAAGTGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151430 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCL2 - BCL2, apoptosis regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000633.2 | 5118 | UTR 3 | NP_000624.2 | |||
NM_000657.2 | 5118 | Intron | NP_000648.2 | |||
XM_011526135.2 | 5118 | Intron | XP_011524437.1 | |||
XM_017025917.1 | 5118 | Intron | XP_016881406.1 |