Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTTGGTAGGTGTGAGGACATTC[C/T]GAAACACGGCCATCCACATTCTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136351 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FLT3 - fms related tyrosine kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004119.2 | 2873 | Silent Mutation | TCA,TCG | S,S 963 | NP_004110.2 | |
XM_011535015.2 | 2873 | Silent Mutation | TCA,TCG | S,S 944 | XP_011533317.1 | |
XM_011535017.2 | 2873 | Silent Mutation | TCA,TCG | S,S 788 | XP_011533319.1 | |
XM_011535018.2 | 2873 | Silent Mutation | TCA,TCG | S,S 788 | XP_011533320.1 | |
XM_017020486.1 | 2873 | Silent Mutation | TCA,TCG | S,S 891 | XP_016875975.1 | |
XM_017020487.1 | 2873 | Silent Mutation | TCA,TCG | S,S 788 | XP_016875976.1 | |
XM_017020488.1 | 2873 | Silent Mutation | TCA,TCG | S,S 670 | XP_016875977.1 | |
XM_017020489.1 | 2873 | Silent Mutation | TCA,TCG | S,S 664 | XP_016875978.1 |