Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGGATTATACTCCTACTTGGAAA[G/T]GAGCTGAAGACTCATCCTTCAACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604719 MIM: 191110 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLB1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLB1L - galactosidase beta 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STK16 - serine/threonine kinase 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011512067.2 | 2457 | UTR 3 | XP_011510369.1 | |||
XM_017005160.1 | 2457 | Intron | XP_016860649.1 |
TUBA4A - tubulin alpha 4a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278552.1 | 2457 | UTR 3 | NP_001265481.1 | |||
NM_006000.2 | 2457 | UTR 3 | NP_005991.1 | |||
XM_017004824.1 | 2457 | Intron | XP_016860313.1 |
TUBA4B - tubulin alpha 4b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |