Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTACTATGTGACTGAAAATGACTT[G/T]CTCTACTAGATCTGGAACCTGTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616284 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLG-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FLG-AS1 - FLG antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FLG2 - filaggrin family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014342.2 | 6613 | Missense Mutation | GAA,GCA | E,A 2325 | NP_001014364.1 | |
XM_011509531.2 | 6613 | Missense Mutation | GAA,GCA | E,A 2173 | XP_011507833.1 |