Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCCGCGGCCCCGCCCGCGCCGGT[G/T]GAGGACCTGTCCAAAGTGTCGGACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC85A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC85A - coiled-coil domain containing 85A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080433.1 | 601 | Silent Mutation | GTG,GTT | V,V 33 | NP_001073902.1 | |
XM_005264125.1 | 601 | Silent Mutation | GTG,GTT | V,V 33 | XP_005264182.1 |
LOC100129434 - uncharacterized LOC100129434 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105374690 - uncharacterized LOC105374690 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107985795 - uncharacterized LOC107985795 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017005442.1 | 601 | Intron | XP_016860931.1 |