Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGCCCCTGTTATGGGTCAGCCT[C/T]TCCGGGATGGTGGCCGCCAACACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608682 MIM: 606774 MIM: 603560 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADM2 - adrenomedullin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253845.1 | 295 | Missense Mutation | CTC,TTC | L,F 82 | NP_001240774.1 | |
XM_017028940.1 | 295 | Missense Mutation | CTC,TTC | L,F 82 | XP_016884429.1 |
MIOX - myo-inositol oxygenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SBF1 - SET binding factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |