Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCTTGGGCTGCCACACGCATGTG[G/T]ATCAGATGAATGGACATTTTAGTAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
HIGD1A PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
HIGD1A - HIG1 hypoxia inducible domain family member 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099668.1 | 305 | Silent Mutation | ATA,ATC | I,I 75 | NP_001093138.1 | |
NM_001099669.1 | 305 | Silent Mutation | ATA,ATC | I,I 61 | NP_001093139.1 | |
NM_014056.3 | 305 | Silent Mutation | ATA,ATC | I,I 61 | NP_054775.2 |