Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTGGAGCTAAGCGGCCCCGGAC[C/T]ACCATCACAGCCAAGCAGCTGGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616352 MIM: 602146 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACBD6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACBD6 - acyl-CoA binding domain containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032360.3 | 952 | Intron | NP_115736.1 |
LHX4 - LIM homeobox 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033343.3 | 952 | Silent Mutation | ACC,ACT | T,T 162 | NP_203129.1 | |
XM_011510105.2 | 952 | Silent Mutation | ACC,ACT | T,T 101 | XP_011508407.1 | |
XM_011510106.2 | 952 | Silent Mutation | ACC,ACT | T,T 101 | XP_011508408.1 | |
XM_011510108.2 | 952 | Silent Mutation | ACC,ACT | T,T 87 | XP_011508410.1 | |
XM_017002755.1 | 952 | Silent Mutation | ACC,ACT | T,T 87 | XP_016858244.1 |
LHX4-AS1 - LHX4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |