Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTAGGAATAGACCCTGGCGTCCAC[C/T]GTGAGATGTTCTTCAGCTACCAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609230 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDT5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUDT5 - nudix hydrolase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321647.1 | 992 | Silent Mutation | NP_001308576.1 | |||
NM_001321648.1 | 992 | Silent Mutation | NP_001308577.1 | |||
NM_014142.3 | 992 | Silent Mutation | NP_054861.2 |
SEC61A2 - Sec61 translocon alpha 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142627.2 | 992 | Intron | NP_001136099.1 | |||
NM_001142628.1 | 992 | Intron | NP_001136100.1 | |||
NM_018144.3 | 992 | Intron | NP_060614.2 |