Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGACTCTGGAAGAGCCAGGCGAG[C/T]ACCCACGAGATGGGGCACGGCGGGC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
EFCC1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
EFCC1 - EF-hand and coiled-coil domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024768.2 | 180 | Silent Mutation | AGC,AGT | S,S 241 | NP_079044.2 | |
XM_005247781.3 | 180 | Silent Mutation | AGC,AGT | S,S 241 | XP_005247838.1 | |
XM_011513161.2 | 180 | Silent Mutation | AGC,AGT | S,S 12 | XP_011511463.1 | |
XM_011513163.2 | 180 | Silent Mutation | AGC,AGT | S,S 241 | XP_011511465.1 | |
XM_011513164.2 | 180 | Intron | XP_011511466.1 |
KIAA1257 - KIAA1257 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020741.2 | 180 | Intron | NP_065792.1 | |||
XM_011513050.2 | 180 | Intron | XP_011511352.2 | |||
XM_017006939.1 | 180 | Intron | XP_016862428.1 | |||
XM_017006940.1 | 180 | Intron | XP_016862429.1 | |||
XM_017006941.1 | 180 | Intron | XP_016862430.1 | |||
XM_017006942.1 | 180 | Intron | XP_016862431.1 | |||
XM_017006943.1 | 180 | Intron | XP_016862432.1 | |||
XM_017006944.1 | 180 | Intron | XP_016862433.1 | |||
XM_017006945.1 | 180 | Intron | XP_016862434.1 | |||
XM_017006946.1 | 180 | Intron | XP_016862435.1 | |||
XM_017006947.1 | 180 | Intron | XP_016862436.1 | |||
XM_017006948.1 | 180 | Intron | XP_016862437.1 | |||
XM_017006949.1 | 180 | Intron | XP_016862438.1 | |||
XM_017006950.1 | 180 | Intron | XP_016862439.1 | |||
XM_017006951.1 | 180 | Intron | XP_016862440.1 | |||
XM_017006952.1 | 180 | Intron | XP_016862441.1 | |||
XM_017006953.1 | 180 | Intron | XP_016862442.1 | |||
XM_017006954.1 | 180 | Intron | XP_016862443.1 |