Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCCACTTTCCCTAGAAAGAGAA[C/T]ACCGTCTGTGGCTTTATTTGTAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615677 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERPINA9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SERPINA9 - serpin family A member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042518.1 | 1344 | Missense Mutation | ATT,GTT | I,V 323 | NP_001035983.1 | |
NM_001284275.1 | 1344 | Missense Mutation | ATT,GTT | I,V 387 | NP_001271204.1 | |
NM_001284276.1 | 1344 | Missense Mutation | ATT,GTT | I,V 274 | NP_001271205.1 | |
NM_175739.3 | 1344 | Missense Mutation | ATT,GTT | I,V 423 | NP_783866.2 | |
XM_011536714.2 | 1344 | Missense Mutation | ATT,GTT | I,V 405 | XP_011535016.1 | |
XM_011536715.2 | 1344 | Missense Mutation | ATT,GTT | I,V 405 | XP_011535017.1 | |
XM_011536716.2 | 1344 | Missense Mutation | ATT,GTT | I,V 325 | XP_011535018.1 | |
XM_011536717.1 | 1344 | Missense Mutation | ATT,GTT | I,V 325 | XP_011535019.1 |