Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCAGGTAGGCGGCGTCGTCGCGC[A/G]GGCTGCGCTCATTGTCGCCGCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608234 MIM: 605591 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAL3ST3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GAL3ST3 - galactose-3-O-sulfotransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033036.2 | 846 | Missense Mutation | NP_149025.1 | |||
XM_017018519.1 | 846 | Missense Mutation | XP_016874008.1 |
SF3B2 - splicing factor 3b subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |