Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCCGGCTCCGGCTCGGTGATGCT[C/T]CCCGCGGGGATGATTAACCCTTCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604889 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NBEA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NBEA - neurobeachin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204197.1 | 702 | Intron | NP_001191126.1 | |||
NM_015678.4 | 702 | Silent Mutation | CTC,CTT | L,L 56 | NP_056493.3 | |
XM_005266346.4 | 702 | Silent Mutation | CTC,CTT | L,L 56 | XP_005266403.1 | |
XM_005266347.4 | 702 | Silent Mutation | CTC,CTT | L,L 56 | XP_005266404.2 | |
XM_005266348.4 | 702 | Silent Mutation | CTC,CTT | L,L 56 | XP_005266405.2 | |
XM_006719803.3 | 702 | Silent Mutation | CTC,CTT | L,L 56 | XP_006719866.1 | |
XM_006719805.3 | 702 | Silent Mutation | CTC,CTT | L,L 56 | XP_006719868.1 | |
XM_006719806.3 | 702 | Silent Mutation | CTC,CTT | L,L 56 | XP_006719869.1 | |
XM_011535046.2 | 702 | Intron | XP_011533348.1 | |||
XM_011535047.2 | 702 | Silent Mutation | CTC,CTT | L,L 56 | XP_011533349.1 | |
XM_017020544.1 | 702 | Silent Mutation | CTC,CTT | L,L 56 | XP_016876033.1 | |
XM_017020545.1 | 702 | Silent Mutation | CTC,CTT | L,L 56 | XP_016876034.1 | |
XM_017020546.1 | 702 | Intron | XP_016876035.1 |