Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCCTGGGCGTCATCCCCAACAC[A/G]CCTGACCCCGCCAGCGACGCGCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607040 | ||||||||||||||||||||
Literature Links: |
ABCC11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCC11 - ATP binding cassette subfamily C member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032583.3 | 411 | Intron | NP_115972.2 | |||
NM_033151.3 | 411 | Intron | NP_149163.2 | |||
NM_145186.2 | 411 | Intron | NP_660187.1 | |||
XM_011523397.1 | 411 | Intron | XP_011521699.1 | |||
XM_011523398.2 | 411 | Intron | XP_011521700.1 | |||
XM_017023795.1 | 411 | Intron | XP_016879284.1 | |||
XM_017023796.1 | 411 | Intron | XP_016879285.1 | |||
XM_017023797.1 | 411 | Intron | XP_016879286.1 | |||
XM_017023798.1 | 411 | Intron | XP_016879287.1 | |||
XM_017023799.1 | 411 | Intron | XP_016879288.1 | |||
XM_017023800.1 | 411 | Intron | XP_016879289.1 | |||
XM_017023801.1 | 411 | Intron | XP_016879290.1 | |||
XM_017023802.1 | 411 | Intron | XP_016879291.1 | |||
XM_017023803.1 | 411 | Intron | XP_016879292.1 |
LONP2 - lon peptidase 2, peroxisomal | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300948.1 | 411 | Silent Mutation | ACA,ACG | T,T 63 | NP_001287877.1 | |
NM_031490.3 | 411 | Silent Mutation | ACA,ACG | T,T 63 | NP_113678.2 | |
XM_017023755.1 | 411 | Silent Mutation | ACA,ACG | T,T 63 | XP_016879244.1 | |
XM_017023756.1 | 411 | Silent Mutation | ACA,ACG | T,T 63 | XP_016879245.1 |