Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCGTTCTCCAGAGCCTCCCGTTC[C/T]GGCCGGTGGACGACGCCGCAGCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616599 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BORCS6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BORCS6 - BLOC-1 related complex subunit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017622.2 | 325 | Silent Mutation | CCA,CCG | P,P 73 | NP_060092.2 |
MIR4521 - microRNA 4521 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |