Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGATTATCCCTTCATCGTGTAGC[C/T]GCTTCCAGACTTCTCTCTCCAGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610138 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST6GALNAC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ST6GALNAC1 - ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289107.1 | 1933 | Missense Mutation | CAG,CGG | Q,R 446 | NP_001276036.1 | |
NM_018414.4 | 1933 | Missense Mutation | CAG,CGG | Q,R 578 | NP_060884.1 | |
XM_011524995.2 | 1933 | Intron | XP_011523297.1 | |||
XM_011524996.2 | 1933 | Intron | XP_011523298.1 | |||
XM_017024842.1 | 1933 | Intron | XP_016880331.1 | |||
XM_017024843.1 | 1933 | Intron | XP_016880332.1 | |||
XM_017024844.1 | 1933 | Intron | XP_016880333.1 |