Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAATGTGAAATCAACATGGCGGCT[A/G]TGTCTTCTGAGCCCATAACAGATGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 615852 MIM: 616883 | |||||||||||||||||||||||
Literature Links: |
RAB6B PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
RAB6B - RAB6B, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016577.3 | 5142 | UTR 3 | NP_057661.3 | |||
XM_011512893.2 | 5142 | UTR 3 | XP_011511195.1 |
SRPRB - SRP receptor beta subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |