Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATCCTGGGACAACTGGTACCCCA[C/G]ATCCTGGAACAACTGGTACCACACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 154360 | ||||||||||||||||||||
Literature Links: |
MGAM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MGAM - maltase-glucoamylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004668.2 | 368 | Missense Mutation | CAT,GAT | H,D 61 | NP_004659.2 | |
XM_006716168.2 | 368 | Missense Mutation | CAT,GAT | H,D 61 | XP_006716231.1 | |
XM_011516670.2 | 368 | Missense Mutation | CAT,GAT | H,D 61 | XP_011514972.1 | |
XM_011516671.2 | 368 | Missense Mutation | CAT,GAT | H,D 61 | XP_011514973.1 | |
XM_011516672.2 | 368 | Missense Mutation | CAT,GAT | H,D 61 | XP_011514974.1 | |
XM_011516673.2 | 368 | Missense Mutation | CAT,GAT | H,D 61 | XP_011514975.1 | |
XM_011516674.2 | 368 | Missense Mutation | CAT,GAT | H,D 61 | XP_011514976.1 | |
XM_017012772.1 | 368 | Missense Mutation | CAT,GAT | H,D 61 | XP_016868261.1 |