Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATTTTAACAAAAGAAACCTCAG[C/T]TTATAATTTTCTTACCAGGATTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600412 MIM: 612468 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GBP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GBP2 - guanylate binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GBP7 - guanylate binding protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207398.2 | 2052 | Missense Mutation | AAC,AGC | N,S 638 | NP_997281.2 |