Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTTGCCATGAACAGTGGCGGCGG[C/G]CTCCCGCCCCCCTCGGCCGCCGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PAN3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PAN3 - PAN3 poly(A) specific ribonuclease subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175854.7 | 170 | Silent Mutation | GGC,GGG | G,G 6 | NP_787050.6 | |
XM_005266332.3 | 170 | Silent Mutation | GGC,GGG | G,G 6 | XP_005266389.1 | |
XM_005266333.3 | 170 | Silent Mutation | GGC,GGG | G,G 6 | XP_005266390.1 | |
XM_005266334.1 | 170 | Intron | XP_005266391.1 | |||
XM_011535032.2 | 170 | Silent Mutation | GGC,GGG | G,G 6 | XP_011533334.1 | |
XM_011535033.2 | 170 | Silent Mutation | GGC,GGG | G,G 6 | XP_011533335.1 | |
XM_011535034.2 | 170 | Intron | XP_011533336.1 | |||
XM_017020528.1 | 170 | Silent Mutation | GGC,GGG | G,G 6 | XP_016876017.1 | |
XM_017020529.1 | 170 | Intron | XP_016876018.1 | |||
XM_017020530.1 | 170 | Silent Mutation | GGC,GGG | G,G 6 | XP_016876019.1 | |
XM_017020531.1 | 170 | Intron | XP_016876020.1 | |||
XM_017020532.1 | 170 | Intron | XP_016876021.1 | |||
XM_017020533.1 | 170 | UTR 5 | XP_016876022.1 |
PAN3-AS1 - PAN3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |