Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGAAGTCGTCCTGAACCTCTTCA[A/G]CTGGTAAGCGGGCACCGCCAGGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300349 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRQ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GABRQ - gamma-aminobutyric acid type A receptor theta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018558.3 | 166 | Missense Mutation | AAC,AGC | N,S 49 | NP_061028.3 | |
XM_011531184.2 | 166 | Missense Mutation | AAC,AGC | N,S 49 | XP_011529486.1 |