Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCAGCTCCAGAATGCTATTCAG[G/T]CAGGCATCCTAGCTGAGAAAGATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300941 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC286437 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC286437 - uncharacterized LOC286437 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A53 - solute carrier family 25 member 53 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012755.4 | 1690 | Intron | NP_001012773.2 | |||
XM_005262129.4 | 1690 | Intron | XP_005262186.1 | |||
XM_011530952.2 | 1690 | Intron | XP_011529254.1 | |||
XM_011530953.2 | 1690 | Intron | XP_011529255.1 | |||
XM_017029514.1 | 1690 | Intron | XP_016885003.1 | |||
XM_017029515.1 | 1690 | Intron | XP_016885004.1 | |||
XM_017029516.1 | 1690 | Intron | XP_016885005.1 |
ZCCHC18 - zinc finger CCHC-type containing 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143978.2 | 1690 | Missense Mutation | GCA,TCA | A,S 162 | NP_001137450.1 | |
XM_011531012.2 | 1690 | Missense Mutation | GCA,TCA | A,S 162 | XP_011529314.1 |