Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCTGGGCCAAGGAGACAGGGAA[A/G]TACACCGAAGGCGTGGATGAAGCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607218 MIM: 610032 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IRF5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IRF5 - interferon regulatory factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098627.3 | 457 | Silent Mutation | AAA,AAG | K,K 73 | NP_001092097.2 | |
NM_001098629.2 | 457 | Silent Mutation | AAA,AAG | K,K 73 | NP_001092099.1 | |
NM_001098630.2 | 457 | Silent Mutation | AAA,AAG | K,K 73 | NP_001092100.1 | |
NM_001242452.2 | 457 | Silent Mutation | AAA,AAG | K,K 73 | NP_001229381.1 | |
NM_032643.4 | 457 | Silent Mutation | AAA,AAG | K,K 73 | NP_116032.1 | |
XM_005250317.3 | 457 | Silent Mutation | AAA,AAG | K,K 73 | XP_005250374.1 | |
XM_006715974.2 | 457 | Silent Mutation | AAA,AAG | K,K 73 | XP_006716037.1 | |
XM_011516158.2 | 457 | Silent Mutation | AAA,AAG | K,K 73 | XP_011514460.1 | |
XM_011516159.2 | 457 | Silent Mutation | AAA,AAG | K,K 73 | XP_011514461.1 | |
XM_011516160.1 | 457 | Silent Mutation | AAA,AAG | K,K 73 | XP_011514462.1 | |
XM_011516161.1 | 457 | Silent Mutation | AAA,AAG | K,K 73 | XP_011514463.1 | |
XM_011516162.1 | 457 | Silent Mutation | AAA,AAG | K,K 73 | XP_011514464.1 | |
XM_011516163.2 | 457 | Silent Mutation | AAA,AAG | K,K 73 | XP_011514465.1 | |
XM_011516164.1 | 457 | Silent Mutation | AAA,AAG | K,K 73 | XP_011514466.1 |
TNPO3 - transportin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |